Learn More
Mouse CD160 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marque: R&D Systems AF3899
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD160 Polyclonal specifically detects CD160 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Spécification
| CD160 | |
| Polyclonal | |
| Unconjugated | |
| AAH21596 | |
| CD160 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD160 Gly28-Ser160 Accession # AAH21596 | |
| 100 μg | |
| Primary | |
| Detects mouse CD160 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human CD160 is observed. | |
| Mouse | |
| IgG |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| BY55, BY55FLJ46513, CD160, CD160 antigenimmunoglobulin superfamily member, CD160 molecule, Natural killer cell receptor BY55, NK1, NK28 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 11126 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |