missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CEACAM-1/CD66a Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Marque: R&D Systems FAB6480R-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Mouse samples. It is validated for Flow Cytometry.
Spécification
| CEACAM1/CD66a | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P31809 | |
| CEACAM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-1/CD66a Glu35-Gly428 Accession # P31809 | |
| RUO | |
| 634 | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| 723629 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| antigen CD66, BGP1, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, Biliary Glycoprotein 1, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen, Cea-1, CEACAM1, Hv-1, Hv-2, MHVR | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse CEACAM-1/CD66a in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CECAM-1, -3, -5, or -6 is observed. In Western blots, no cross-reactivity with rhCEACAM-1, -3, -4, -5, -6, or -7 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only (RUO)
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu