missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CX3CL1/Fractalkine Chemokine Domain Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Marque: R&D Systems MAB571-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Spécification
| CX3CL1/Fractalkine | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CX3CL1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CX3CL1/Fractalkine, S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse CX3CL1/Fractalkine, and E. coli-derived recombinant mouse CX3CL1/Fractalkine | |
| 25 μg | |
| Primary | |
| Detects mouse CX3CL1/Fractalkine in Western blots. In direct ELISAs, this antibody shows 100% cross-reactivity with the chemokine domain of recombinant human (rh) CX3CL1, 13% cross-reactivity with rhCX3CL1, and 50% cross-reactivity with recombinant rat CX3CL1. | |
| Mouse | |
| Purified |
| Western Blot, Neutralization, ELISA | |
| 126315 | |
| Western Blot 1 ug/mL, Neutralization 0.75-3.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6376 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu