missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL7/Thymus Chemokine-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Marque: R&D Systems AF793
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CXCL7/Thymus Chemokine-1 Polyclonal specifically detects CXCL7/Thymus Chemokine-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Spécification
| CXCL7/Thymus Chemokine-1 | |
| Polyclonal | |
| Unconjugated | |
| AAG36786 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5473 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.8-4 ug/mL | |
| Ppbp | |
| E. coli-derived recombinant mouse CXCL7/Thymus Chemokine-1 Lys40-Tyr113 Accession # AAG36786 | |
| 100 μg | |
| Primary | |
| Detects CXCL7/Thymus Chemokine-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant rat CXCL7/Thymus Chemokine-1 is observed. | |
| Mouse | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu