missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DEC-205/CD205 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marque: R&D Systems AF5975-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DEC-205/CD205 Polyclonal specifically detects DEC-205/CD205 in Mouse samples. It is validated for Western Blot.
Spécification
| DEC-205/CD205 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LY75 | |
| E. coli-derived recombinant mouse DEC-205/CD205 Cys216-Pro503 Accession # Q60767 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q60767 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Detects mouse DEC-205/CD205 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human DEC-205 is observed. | |
| Mouse | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu