missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-alpha 2/IFNA2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Marque: R&D Systems MAB101491-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IFN-alpha 2 Monoclonal antibody specifically detects IFN-alpha 2 in Mouse samples. It is validated for ELISA
Spécification
| IFN-alpha 2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| alpha-2a interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 | |
| Human embryonic kidney cell, HEK293-derived mouse IFN-alpha 2/IFNA2, Cys24-Glu190, Accession # P01573 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 996548 | |
| ELISA | |
| P01573 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3440 | |
| Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu