missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LAP (TGF-beta 1) Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
Marque: R&D Systems MAB7666
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Spécification
| LAP (TGF-beta 1) | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CED, DPD1, LAP, LAP (TGFbeta 1), TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, transforming growth factor beta 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7040 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 860206 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P04202 | |
| TGFB1 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse LAP (TGF-beta 1) Met1-Ser390 Accession # P04202 | |
| 100 μg | |
| Primary | |
| Detects mouse LAP (TGF-beta 1) in flow cytometry. | |
| Mouse | |
| Supernatant |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu