missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SPARC-like 1/SPARCL1 Antibody, R&D Systems™
Rat Monoclonal Antibody
Marque: R&D Systems MAB2836
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SPARC-like 1/SPARCL1 Monoclonal specifically detects SPARC-like 1/SPARCL1 in Mouse samples. It is validated for Western Blot.
Spécification
| SPARC-like 1/SPARCL1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P70663 | |
| SPARCL1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SPARC-like 1/SPARCL1 Ile17-Phe650 Accession # P70663 | |
| 100 μg | |
| Primary | |
| Detects mouse SPARC-like 1/SPARCL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse SPARC or recombinant human SPARC-like 1 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 348905 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hevin, High endothelial venule protein, MAST 9, MAST9, PIG33, proliferation-inducing protein 33, SC1, SPARC like 1, SPARCL1, SPARC-like 1 (hevin), SPARC-like 1 (mast9, hevin), SPARC-like protein 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8404 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu