missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TREML4/TLT-4 Antibody, R&D Systems™
Goat Polyclonal Antibody
Marque: R&D Systems AF5500
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TREML4/TLT-4 Polyclonal specifically detects TREML4/TLT-4 in Mouse samples. It is validated for Western Blot.
Spécification
| TREML4/TLT-4 | |
| Polyclonal | |
| Unconjugated | |
| NP_001029094 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 285852 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Treml4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREML4/TLT-4 Ser25-Val195 Accession # NP_001029094 | |
| 100 μg | |
| Primary | |
| Detects mouse TREML4/TLT-4 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) TREM1, rmTREM2b, and rmTREM3 is observed. | |
| Mouse | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu