missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse VE-Cadherin Antibody, R&D Systems™
Goat Polyclonal Antibody
Marque: R&D Systems AF1002
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
VE-Cadherin Polyclonal specifically detects VE-Cadherin in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Spécification
| VE-Cadherin | |
| Polyclonal | |
| Unconjugated | |
| 2208309A | |
| CDH5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse VE-Cadherin Asp46-Gln592 Accession # 2208309A | |
| 100 μg | |
| Primary | |
| Detects mouse VE-Cadherin in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human VE-Cadherin is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| 7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1003 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu