Learn More
MP Biomedicals™ Albumin 35% Solution
Globulin free albumins are suitable for use in applications where no other proteins should be present
Marque: MP Biomedicals™ 08810063
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 1.00000kg
Description
Albumins are a group of acidic proteins which occur plentifully in the body fluids and tissues of mammals and in some plant seeds.
- CAS Number: 9048-46-8
- MDL Number: MFCD00130384
- Format: Solution
- Sterile albumins
- Concentration: About 35g/dl
For immunoassays and mammalian cell culture. Used to solubilize lipids and is also used as a blocking agent in Western blots or ELISA applications.
Spécification
34.4g/dL | |
∽66 | |
7.2 | |
0° to 5°C |
Immunoassay, Mammalian Cell Culture | |
Bovine Albumin 35% Solution | |
1L | |
Solution |