Learn More
Novus Biologicals™ MSPR/Ron Beta Antibody (Aveo anti-RON) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-28436-100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MSPR/Ron Beta Monoclonal antibody specifically detects MSPR/Ron Beta in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| MSPR/Ron Beta | |
| Monoclonal | |
| Unconjugated | |
| Q04912 | |
| Human | |
| Protein A purified | |
| RUO | |
| 4486 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Aveo anti-RON | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD136, CDw136, Macrophage-stimulating protein receptor beta chain, MSP receptor, NPCA3, p185-Ron, Protein-tyrosine kinase 8, PTK8, Ron | |
| MSPR/ RON/ CD136 | |
| 100 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |