missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MST2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA585196
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Keep as concentrated solution. Predicted reactivity: Mouse (98%), Rat (97%), Zebrafish (88%), Xenopus laevis (95%), Chicken (97%), Rhesus Monkey (99%), Bovine (99%). Positive Control: 293T, A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STK3 (MST2) is a STE kinase family member that acts upstream of the MAP kinase signaling pathways, and is known to be activated by the proapoptotic stimuli staurosporine and FAS ligand. STK3 is a stress-activated, pro-apoptotic kinase which, following caspase-cleavage, enters the nucleus and induces chromatin condensation followed by internucleosomal DNA fragmentation. It is a key component of the Hippo signaling pathway which plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1, which in turn phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. Phosphorylation of YAP1 by LATS2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration. STK3/MST2 and STK4/MST1 are required to repress proliferation of mature hepatocytes, to prevent activation of facultative adult liver stem cells (oval cells), and to inhibit tumor formation.
Spécification
| MST2 | |
| Polyclonal | |
| Unconjugated | |
| STK3 | |
| 0610042I06Rik; KB-1458E12.1; KRS1; Mammalian STE20-like protein kinase 2; mess1; MST; Mst2; MST-2; MST2/C; MST2/N; Mst3; protein kinase homolog; serine/threonine kinase 3; serine/threonine kinase 3 (STE20 homolog, yeast); serine/threonine kinase 3 (Ste20, yeast homolog); serine/threonine-protein kinase 3; Serine/threonine-protein kinase 3 20kDa subunit; Serine/threonine-protein kinase 3 36kDa subunit; Serine/threonine-protein kinase Krs-1; Ste20; STE20-like kinase MST2; STK3; wu:fc19e11; zgc:55383 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6788 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.8 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q13188 | |
| STK3 | |
| Recombinant protein encompassing a sequence within the center region of human STK3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu