missing translation for 'onlineSavingsMsg'
Learn More

MUC1 Antibody (MUC1/967), PE/Cy5.5, Novus Biologicals™

Code produit 18132459 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18132459 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18132459 Fournisseur Novus Biologicals Code fournisseur NBP247885PECY55

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

MUC1 Monoclonal specifically detects MUC1 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Spécification

Antigène MUC1
Applications Flow Cytometry
Classification Monoclonal
Clone MUC1/967
Conjugué PE-Cyanine5.5
Dilution Flow Cytometry
Formule PBS with 0.05% Sodium Azide
Alias de gène Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, CD227, CD227 antigen, DF3 antigen, EMA, episialin, H23 antigen, H23AG, KL-6, MAM6, MUC-1, MUC1/ZD, mucin 1, cell surface associated, mucin 1, transmembrane, mucin-1, Peanut-reactive urinary mucin, PEMMUC-1/SEC, PEMT, Polymorphic epithelial mucin, PUMMUC-1/X, tumor associated epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin
Symboles de gène(s) MUC1
Espèces hôtes Mouse
Immunogène Human milk-fat globule membranes (HMFGM) (Uniprot: P15941 )
Méthode de purification Protein A or G purified
Quantité 0.1 mL
Disciplines de recherche Cancer, Cellular Markers, Extracellular Matrix, Inflammation, Signal Transduction
Primaire ou secondaire Primary
Identification génétique (Entrez) 4582.0
Spécificité du test This monoclonal antibody recognizes full-length MUC1 in a glycosylation-independent manner and can bind to the fully glycosylated protein. The dominant epitope of this monoclonal antibody is APDTR in the VNTR region. It reacts with the core peptide of the MUC1 protein, which is a member of a family of mucin glycoproteins that are characterized by high carbohydrate content, O-linked oligosaccharides, high molecular weight (200kDa) and an amino acid composition rich in serine, threonine, proline and glycine. The core protein contains a domain of 20 amino-acid tandem repeats that functions as multiple epitopes for the monoclonal antibody. Incomplete glycosylation of some tumor-associated mucins may lead to variable unmasking of the multiple peptide epitopes leading to the observed differences in staining intensity between normal and malignant tissues. This monoclonal antibody reacts with both normal and malignant epithelia of various tissues including breast and colon.
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.