missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MuRF1/TRIM63 Monoclonal antibody specifically detects MuRF1/TRIM63 in Human, Mouse, Rat samples. It is validated for Western Blot
Spécification
Spécification
| Antigène | MuRF1/TRIM63 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | M316 |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1:250 |
| Formule | PBS, 1 mg/mL BSA and 50% Glycerol |
| Alias de gène | EC 6.3.2.-, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28FLJ32380, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZMURF-1, Striated muscle RING zinc finger protein, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63 |
| Espèces hôtes | Mouse |
| Immunogène | Clone M316 was generated from a synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal half of human MuRF1/TRIM63. This peptide sequence is highly conserved in rat and mouse MuRF1/TRIM63, and has 50% homology to MuRF2 (TRIM-55). [UniProt# Q969Q1] |
| Afficher plus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?