missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MxA/Mx1 Antibody (OTI2G12), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Marque: Novus Biologicals NBP1-47859
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MxA/Mx1 Monoclonal specifically detects MxA/Mx1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Spécification
| MxA/Mx1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| MX1 | |
| Full length human recombinant protein of human MX1 (NP_002453) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for Homo sapiens myxovirus (influenza virus) resistance 1, interferon-inducible protein p78 (mouse) (MX1), transcript variant 2. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI2G12 | |
| Unconjugated | |
| human interferon-regulated resistance GTP-binding protein MXA10IFI-78Khomolog of murine (interferon-inducibleprotein p78), myxovirus (influenza virus) resistance 1, interferon-inducible protein p78(mouse) | |
| Mouse | |
| 75.5 kDa | |
| 0.1 mL | |
| Apoptosis | |
| 4599 | |
| Human, Mouse, Monkey | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu