missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MYLK4, GST Tag Recombinant Protein
This MYLK4 Recombinant Human Protein (full length) was expressed in insect cells.
Marque: Invitrogen™ A32898
Informations supplémentaires : Poids : 0.00900kg
Description
MYLK4 serine/threonine kinase or myosin light chain kinase family member 4 is a member of the CAMK kinase family. It is one of four myosin light chain kinases noted in a comprehensive analysis of protein kinases encoded by the human genome.- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Spécification
AAI32834 | |
70.9 kDa | |
CAMK Unique Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Full Length, Recombinant, Active |
Kinase Assay | |
100 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Insect | |
Liquid |