missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myosin Phosphatase, p Thr853 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-13185
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Myosin Phosphatase (p Thr853) Polyclonal antibody specifically detects Myosin Phosphatase (p Thr853) in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot.
Spécification
| Myosin Phosphatase (p Thr853) | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH 7.4), 0.15M NaCl and 30% Glycerol with 0.05% Sodium Azide | |
| MBSmyosin phosphatase, target subunit 1, MGC133042, Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, MYPT1protein phosphatase 1 regulatory subunit 12A, protein phosphatase 1, regulatory (inhibitor) subunit 12A, Protein phosphatase myosin-binding subunit | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes recombinant truncated MYPT1 phosphorylated on Thr853, MW ∼80kDa | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-2 ug/ml | |
| Q90623 | |
| PPP1R12A | |
| Synthetic peptide corresponding to aa845-855, C-EKRRS[pT]GVSFW of chicken MYPT1 with an N-terminal cysteine added for conjugation purposes (Q90623). Phosphorylation site corresponds to Thr853 in human MYPT1. | |
| 0.2 mg | |
| Phospho Specific, Protein Phosphatase | |
| 4659 | |
| Rat | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu