Learn More
Invitrogen™ Myostatin Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5114224
Description
Immunogen sequence: APKRYKANYC SGECEFVFLQ KYPHTHLVHQ ANPRGSAGPC CTPTKMSPIN For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%;.
The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.
Spécification
| Myostatin | |
| Polyclonal | |
| Unconjugated | |
| Mstn | |
| Cmpt; compact; Gdf8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; Myostatin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17700 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O08689 | |
| Mstn | |
| Synthetic peptide directed towards the C-terminal region of Mouse Mstn (aa 301-350). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.