missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Myostatin Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA511936
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is predicted to react with bovine, porcine and non-human primate based on sequence homology.
The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.
Spécification
| Myostatin | |
| Polyclonal | |
| Unconjugated | |
| Mstn | |
| Cmpt; compact; Gdf8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; Myostatin | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 17700, 2660 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O08689, O14793 | |
| Mstn | |
| KLH conjugated synthetic peptide between 9-38 amino acids from the N-terminal region of human Myostatin (GDF8). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu