Learn More
MYST3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Marque:  Novus Biologicals NBP1-44049
Code nomenclature Nacres: NA.46
Code produit. 18716233
  • 429.00€ / 0.10mg
Description
MYST3 Polyclonal specifically detects MYST3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
MYST3
Western Blot 1 ug/ml, Immunohistochemistry 3.75 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml, Peptide ELISA Detection limit 1:16000
Goat
RUO
IgG
Western Blot
EC 2.3.1.48, EC 3.1.26.4, histone acetyltransferase MYST3, KAT6A, MGC167033, MOZMonocytic leukemia zinc finger protein, MYST histone acetyltransferase (monocytic leukemia) 3, MYST-3, runt-related transcription factor binding protein 2, Runt-related transcription factor-binding protein 2, RUNXBP2, Zinc finger protein 220, ZNF220MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3
Affinity Purified
Store at -20C. Avoid freeze-thaw cycles.
For Research Use Only