missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Myc Antibody (NMYC-1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Marque: Novus Biologicals NB200-109
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
n-Myc Monoclonal specifically detects n-Myc in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays, CyTOF-ready.
Spécification
| n-Myc | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500 - 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 21068393), Flow Cytometry reported in scientific literature (PMID 27539729), Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 1:10 - 1:100, Gel Super Shift Assays 1:1 - 1:100. Use reported in scientific literature (PMID 21068393), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready | |
| BHLHE37, bHLHe37N-myc proto-oncogene protein, Class E basic helix-loop-helix protein 37, MODED, neuroblastoma-derived v-myc avian myelocytomatosis viral related oncogene, N-myc, NMYCneuroblastoma MYC oncogene, ODED, oncogene NMYC, pp65/67, v-myc avian myelocytomatosis viral related oncogene, neuroblastoma derived, v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody does not react with c-Myc or L-Myc. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| NMYC-1 | |
| Unconjugated | |
| P04198 | |
| MYCN | |
| A synthetic peptide (SPYVESEDAPPQKC) conjugated to KLH, corresponding to amino acids 327-339 of human n-Myc. [UniProt# P04198] | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors | |
| 4613 | |
| Human, Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu