Learn More
Invitrogen™ N-sulfated Heparan Sulfate Monoclonal Antibody (HepSS-1), Biotin
GREENER_CHOICE
Mouse Monoclonal Antibody
Marque:  Invitrogen™ MA546186
Code nomenclature Nacres: NA.46
Code produit. 17858935
  • 368.00€ / 100µg
Description
This antibody recognizes N-sulfated heparan sulfate (extracellular antigen) present in many species.
Heparan sulfate (HS) is a glycosaminoglycan that is ubiquitously expressed on cell surfaces and in the extracellular matrix and basement membrane. Each HS molecule is a linear polysaccharide composed of repeating disaccharides of hexuronic acid and d-glucosamine that can exhibit immense structural diversity due to substitution to varying extents with sulfate groups and epimerization of glucuronic acid to iduronic acid, with areas of high sulfation and glucuronic acid epimerization being co-located in hot spots throughout the molecule. HS is structurally related to heparin, an extremely highly sulfated form of HS that is restricted to mast cells. The biosynthesis and modification of HS chains is thought to take place within the endoplasmic reticulum, Golgi apparatus, and trans Golgi network, which in the end produce unique HS chains that are covalently attached to a range of core proteins to form HS-proteoglycans [PMID: 24391644].
TRUSTED_SUSTAINABILITY
Spécification
Spécification
N-sulfated Heparan Sulfate
Monoclonal
1 mg/mL
TBS with 15mM sodium azide; pH 8
Mouse
Size-exclusion chromatography
RUO
Chemical
Antibody
IgM
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
HepSS-1
Biotin
Heparan sulfate
MethA murine fibrosarcoma.
100 μg
Primary
4°C, do not freeze
Liquid
Chemical