missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NCOR1 Polyclonal antibody specifically detects NCOR1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigène | NCOR1 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formule | PBS (pH 7.2), 40% Glycerol |
| Alias de gène | hN-CoR, KIAA1047N-CoR1, MGC104216, N-CoRhCIT529I10, nuclear receptor corepressor 1, nuclear receptor co-repressor 1, thyroid hormone- and retinoic acid receptor-associated corepressor 1, TRAC1 |
| Espèces hôtes | Rabbit |
| Immunogène | This antibody was developed against a recombinant protein corresponding to amino acids: PPTSTFQNSPSALVSTPVRTKTSNRYSPESQAQSVHHQRPGSRVSPENLVDKSRGSRPGKSPERSHVSSEPYEPISPPQVPVVHEKQDSLLLLSQ |
| Méthode de purification | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?