Learn More
Nebulin Antibody (NB2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
232.00€
Spécification
| Antigène | Nebulin |
|---|---|
| Clone | NB2 |
| Dilution | Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:400 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
18527741
|
Novus Biologicals
NB120-11099-0.025ml |
0.025 mL |
232.00€
25µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Nebulin Monoclonal specifically detects Nebulin in Human, Mouse, Rat, Porcine, Alligator, Amphibian, Avian, Bovine, Chicken, Fish, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Spécification
| Nebulin | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:400 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| DKFZp686C1456, FLJ11505, FLJ36536, FLJ39568, FLJ39584, NEB177D, nebulin, NEM2 | |
| NEB | |
| IgG1 | |
| Unpurified | |
| Localizes nebulin in skeletal muscle of a wide variety of species from cold-blooded vertebrates to man. Does not crossreact with smooth or cardiac muscle, non-muscle tissue and cultured cells. Localizes nebulin within the I band of skeletal muscle sections by indirect immunofluorescence. |
| NB2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cytoskeleton Markers | |
| Ascites with 15mM Sodium Azide | |
| 4703 | |
| Full length native protein (purified) (Chicken). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only