missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAB1 Antibody (CL0580) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-52944F
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NECAB1 Monoclonal antibody specifically detects NECAB1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spécification
| NECAB1 | |
| Monoclonal | |
| FITC | |
| EF hand calcium binding protein 1, EFCBP1, EF-hand calcium binding protein 1, EF-hand calcium-binding protein 1, neuronal calcium binding protein, Neuronal calcium-binding protein 1, N-terminal EF-hand calcium binding protein 1, N-terminal EF-hand calcium-binding protein 1, STIP-1, synaptotagmin interacting protein 1 | |
| This antibody was developed using a recombinant protein derived from Q8N987, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0580 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 64168 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu