Learn More
Neomycin phosphotransferase II Antibody (4B4D1), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB110-60487
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Neomycin phosphotransferase II Monoclonal specifically detects Neomycin phosphotransferase II in Non-species specific, Bacteria samples. It is validated for Western Blot, ELISA.
Spécification
| Neomycin phosphotransferase II | |
| Monoclonal | |
| Unconjugated | |
| NPT2, NPTII | |
| Ni-NTA purified truncated recombinant NPT-II expressed in E. Coli strain BL21 (DE3) | |
| 0.1 mL | |
| Primary | |
| Non-species specific, Bacteria | |
| Ascites |
| Western Blot, ELISA | |
| 4B4D1 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Transfected or transformed cells/organisms (plants, animals) carrying NPTII gene encoding neomycin phosphotransferase II enzyme. Escherichia coli (E. coli). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Correction du contenu d'un produit
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu