Learn More
Invitrogen™ Nestin Monoclonal Antibody (10C2), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1586.00€ - 3585.00€
Spécification
| Antigène | Nestin |
|---|---|
| Clone | 10C2 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugué | Unconjugated |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
17130802
|
Invitrogen™
14-9843-37 |
2 mg |
1586.00€
2mg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
17140802
|
Affymetrix eBioscience
14-9843-95 |
10 mg | |||||||
Description
The MAb 10C2 recognizes human nestin (residues 1464-1614). Nestin is a 220-240 kDa Class VI intermediate filament protein that is expressed in stem cells of the developing nervous system. Expression has also been found in pancreatic islet and mesenchymal precursors in addition to cells with angiogenic potential. Upon differentiation, nestin-containing filaments are replaced by cell type-specific intermediate filament proteins, such as GFAP. Expression has been identified in a variety of cancerous cells including glioblastomas and pancreatic tumors. No cross-reactivity to rodent nestins has been observed using the 10C2 antibody.
Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. Nestin has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, Nestin is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. Highly specific antibodies to human Nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells. Nestin expression has been observed in pancreatic hematopoietic stem cell populations, and is considered a novel angiogenesis marker of proliferating endothelial cells in colorectal cancer tissue.Spécification
| Nestin | |
| 0.5 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| P48681 | |
| IgG1 | |
| Affinity chromatography | |
| Antibody |
| 10C2 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 10763 | |
| Primary | |
| 4°C | |
| Nes |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.