Learn More
Nestin Mouse, anti-Human, Unconjugated, Clone: 10C2, STEMCELL Technologies
Mouse Monoclonal Antibody
Marque:  Stemcell Technologies 60091.1
Encompass_Preferred
Code nomenclature Nacres: NA.46
Code produit. 17198961
  • 210.60€ / 25µg
Description
  • The 10C2 antibody reacts with human nestin, a class VI intermediate filament (IF) protein expressed by embryonic neuroepithelial stem cells in the developing nervous system and by pancreatic islet and mesenchymal progenitor cells
  • Nestin expression is also observed in glioblastomas and many primary and metastatic melanomas and it is a useful angiogenic marker to evaluate neovascularity of endothelial cells in tumors
  • Nestin is required for the survival and renewal of neural progenitor cells and plays roles in cellular remodeling by forming heterodimers with vimentin or alpha-internexin, a process that appears to be tightly regulated by phosphorylation
  • Nestin localizes to the growth cones during axon elongation in differentiating neurons and may play a role in growth cone guidance
  • Nestin expression is down-regulated during differentiation, typically disappearing by embryonic stage E18
  • The 10C2 antibody reportedly cross-reacts weakly with monkey nestin but not rodent nestin
Spécification
Spécification
Nestin
Monoclonal
Unconjugated
25 μg
Cynomolgus, Human
Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Neurons
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
10C2
Mouse
Primary
IgG1 κ
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.