Learn More
NEURL Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 18898-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The NEURL antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NEURL. This antibody recognizes human, mouse, rat antigen. The NEURL antibody has been validated for the following applications: IP, WB, ELISA analysis.
Spécification
| NEURL | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 9148 | |
| IgG |
| Western Blot, Immunoprecipitation, ELISA | |
| 147 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |