missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurogranin Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-97430PE
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Neurogranin Polyclonal antibody specifically detects Neurogranin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spécification
| Neurogranin | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| calmodulin-binding protein, hng, neurogranin, neurogranin (protein kinase C substrate, RC3), Ng, protein kinase C substrate, RC3ng | |
| Produced in rabbits immunized with E. coli-derived Human Neurogranin fragment. | |
| 0.1 mL | |
| Neuroscience | |
| 4900 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction