missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NF-H Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA316721
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NF-H Polyclonal Antibody for Western Blot, IHC (P), IHC (F), IHC (Free), ICC/IF, ELISA
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
Spécification
| NF-H | |
| Polyclonal | |
| Unconjugated | |
| Nefh | |
| 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig, Horse | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| Whole serum with 0.035% sodium azide | |
| P12036, P12037, P16884, P19246 | |
| Nefh | |
| Purified bovine Neurofilament Heavy protein. | |
| RUO | |
| 100064328, 100156492, 24587, 380684, 4744, 528842 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu