missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-L Antibody (7D1), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-50612
This item is not returnable.
View return policy
Description
NF-L Monoclonal specifically detects NF-L in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| NF-L | |
| Monoclonal | |
| Unconjugated | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| NEFL | |
| A preparation of pig Neurofilament Light and Neurofilament Heavy | |
| 0.1 mL | |
| Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
| 4747 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1 mg/ml | |
| Western Blot 1:5000 - 1:10000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Mammalia | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction