missing translation for 'onlineSavingsMsg'
Learn More

NFIL3 Antibody (1218A), Alexa Fluor™ 700, Novus Biologicals™

Code produit 30064249 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30064249 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30064249 Supplier Novus Biologicals Supplier No. FAB8888N

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NFIL3 Monoclonal antibody specifically detects NFIL3 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigène NFIL3
Applications Flow Cytometry
Classification Monoclonal
Clone 1218A
Conjugué Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry
Formule 50mM Sodium Borate
Alias de gène E4BP4IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A
Espèces hôtes Rabbit
Immunogène E. coli-derived recombinant mouse NFIL3/E4BP4, Lys140-Arg462, Accession # O08750
Méthode de purification Protein A or G purified from cell culture supernatant
Quantité 0.1 mL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 4783
Espèces cibles Mouse
Contenu et stockage Store at 4C in the dark.
Forme Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.