Learn More
NGFI-B alpha/Nur77/NR4A1 Antibody, PE, Novus Biologicals™
Code produit 18506114 Tous les produits Bio Techne Produits
Code produit 18506114 Fournisseur Novus Biologicals Code fournisseur NB10056745PE
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
NGFI-B alpha/Nur77/NR4A1 Polyclonal antibody specifically detects NGFI-B alpha/Nur77/NR4A1 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP)