Learn More
NK1.1 (CD161) Mouse anti-Mouse, APC, Clone: PK136, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech APC-65138-100UG
Description
The NK1.1 (CD161) antibody from Proteintech is a mouse monoclonal antibody generated with nk-1+ cells from mouse spleen and bone marrow of mouse NK1.1 (CD161). This antibody recognizes mouse antigen. The NK1.1 (CD161) antibody has been validated for the following applications: FC analysis. This antibody is apc.
Spécification
| NK1.1 (CD161) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| PK136 | |
| APC | |
| 100 μg | |
| 17057 | |
| IgG2a |