missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Marque: Abnova H00022861-B01P.50ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Sequence: MAGGAWGRLACYLEFLKKEELKEFQLLLANKAHSRSSSGETPAQPEKTSGMEVASYLVAQYGEQRAWDLALHTWEQMGLRSLCAQAQEGAGHSPSFPYSPSEPHLGSPSQPTSTAVLMPWIHELPAGCTQGSERRVLRQLPDTSGRRWREISASLLYQALPSSPDHESPSQESPNAPTSTAVLGSWGSPPQPSLAPREQEAPGTQWPLDETSGIYYTEIREREREKSEKGRPPWAAVVGTPPQAHTSLQPHHHPWEPSVRESLCSTWPWKNEDFNQKFTQLLLLQRPHPRSQDPLVKRSWPDYVEENRGHLIEIRDLFGPGLDTQEPRIVILQGAAGIGKSTLARQVKEAWGRGQLYGDRFQHVFYFSCRELAQSKVVSLAELIGKDGTATPAPIRQILSRPERLLFILDGVDEPGWVLQEPSSELCLHWSQPQPADALLGSLLGKTILPEASFLITARTTALQNLIPSLEQARWVEVLGFSESSRKEYFYRYFTDERQAIRAFRLVKSNKELWALCLVPWVSWLACTCLMQQMKRKEKLTLTSKTTTTLCLHYLAQALQAQPLGPQLRDLCSLAAEGIWQKKTLFSPDDLRKHGLDGAIISTFLKMGILQEHPIPLSYSFIHLCFQEFFAAMSYVLEDEKGRGKHSNCIIDLEKTLEAYGIHGLFGASTTRFLLGLLSDEGEREMENIFHCRLSQGRNLMQWVPSLQLLLQPHSLESLHCLYETRNKTFLTQVMAHFEEMGMCVETDMELLVCTFCIKFSRHVKKLQLIEGRQHRSTWSPTMVVLFRWVPVTDAYWQILFSVLKVTRNLKELDLSGNSLSHSAVKSLCKTLRRPRCLLETLRLAGCGLTAEDCKDLAFGLRANQTLTELDLSFNVLTDAGAKHLCQRLRQPSCKLQRLQLVSCGLTSDCCQDLASVLSASPSLKELDLQQNNLDDVGVRLLCEGLRHPACKLIRLGLDQTTLSDEMRQELRALEQEKPQLLIFSRRKPSVMTPTEGLDTGEMSNSTSSLKRQRLGSERAASHVAQANLKLLDVSKIFPIAEIAEESSPEVVPVELLCVPSPASQGDLHTKPLGTDDDFWGPTGPVATEVVDKEKNLYRVHFPVAGSYRWPNTGLCFVMREAVTVEIEFCVWDQFLGEINPQHSWMVAGPLLDIKAEPGAVEAVHLPHFVALQGGHVDTSLFQMAHFKEEGMLLEKPARVELHHIVLENPSFSPLGVLLKMIHNALRFIPVTSVVLLYHRVHPEEVTFHLYLIPSDCSIRKELELCYRSPGEDQLFSEFYVGHLGSGIRLQVKDKKDETLVWEALVKPGDLMPATTLIPPARIAVPSPLDAPQLLHFVDQYREQLIARVTSVEVVLDKLHGQVLSQEQYERVLAENTRPSQMRKLFSLSQSWDRKCKDGLYQALKETHPHLIMELWEKGSKKGLLPLSSSpécification
| NLRP1 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_014922 | |
| NLRP1 | |
| NLRP1 (NP_055737.1, 1 a.a. ∼ 1429 a.a) full-length human protein. | |
| RUO | |
| 22861 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NLR family, pyrin domain containing 1 | |
| CARD7/CLR17.1/DEFCAP/DEFCAP-L/S/DKFZp586O1822/KIAA0926/NAC/NALP1/PP1044/SLEV1/VAMAS1 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu