missing translation for 'onlineSavingsMsg'
Learn More
Learn More
non-muscle heavy chain 10 Myosin Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals (Bio-Techne) NBP1-18932CL1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
non-muscle heavy chain 10 Myosin Polyclonal antibody specifically detects non-muscle heavy chain 10 Myosin in Human,Mouse samples. It is validated for Western Blot,Immunoprecipitation (Negative)
Spécification
| non-muscle heavy chain 10 Myosin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4628 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Western Blot, Immunoprecipitation (Negative) | |
| CoraFluor 1 | |
| Cellular myosin heavy chain, type B, cellular myosin heavy chain, type B type B, heavy polypeptide 10, non-muscle, MGC134913, MGC134914, Myosin heavy chain 10, Myosin heavy chain, non-muscle IIb, myosin heavy chain, nonmuscle type B, myosin, heavy chain 10, non-muscle, myosin-10, near to the ATP binding region, NMMHC II-b, NMMHCB, NMMHC-B, NMMHC-IIB, Non-muscle myosin heavy chain B, nonmuscle myosin heavy chain IIB, Non-muscle myosin heavy chain IIb, nonmuscle myosin heavy chain-B, nonmuscle myosin II heavy chain-B | |
| The immunogen recognized by this antibody maps to a region between residue 1955 and 1976 of human myosin, heavy chain 10, non-muscle using the numbering given in entry NP_005955.1 (GeneID 4628). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu