Learn More
Noxa Antibody (69N15C9), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Marque: Novus Biologicals NBP2-25266AF750
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Noxa Monoclonal specifically detects Noxa in Mouse samples. It is validated for Western Blot.
Spécification
| Noxa | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PMAIP1 | |
| This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa. | |
| Protein G purified | |
| Apoptosis, Cancer, Cellular Markers, Membrane Trafficking and Chaperones, Necroptosis, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
| 5366.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| 69N15C9 | |
| Western Blot | |
| adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
| Rat | |
| 11.6 kDa | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2b κ |