missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NR1D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA534002
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Percent identity with other species by BLAST analysis: Human (100%); Gorilla, Gibbon, Monkey, Marmoset, Panda, Dog (94%); Bovine, Bat, Horse (88%); Elephant (81%). Antigen retrieval requires Proteinase K treatment.
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
Spécification
| NR1D2 | |
| Polyclonal | |
| Unconjugated | |
| NR1D2 | |
| BD73; EAR-1R; EAR4; HZF2; HZF-2; Nr1d2; nuclear receptor Rev-ErbA beta variant 1; nuclear receptor Rev-ErbA beta variant 2; nuclear receptor subfamily 1 group D member 2; nuclear receptor subfamily 1, group D, member 2; orphan nuclear hormone receptor BD73; orphan nuclear receptor HZF-2; orphan nuclear receptor RVR; orphan receptor; Rev-erb; Rev-erb alpha-related receptor; Rev-erb beta; Rev-erba beta; rev-erba-alpha-related receptor; Rev-erbabeta; Rev-ErbA-beta; rev-erb-beta; Rev-erb-Beta2; RVR; V-erbA-related protein 1-related | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9975 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q14995 | |
| NR1D2 | |
| Synthetic 16 amino acid peptide from N-terminus of human NR1D2. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu