missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NRBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA598977
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NRBP Polyclonal Antibody for Western Blot, IHC (P), ELISA
May play a role in subcellular trafficking between the endoplasmic reticulum and Golgi apparatus through interactions with the Rho-type GTPases. Binding to the NS3 protein of dengue virus type 2 appears to subvert this activity into the alteration of the intracellular membrane structure associated with flaviviral replication. [UniProt].
Spécification
| NRBP | |
| Polyclonal | |
| Unconjugated | |
| NRBP1 | |
| B230344L17Rik; BCON3; HLS7-interacting protein kinase; Madm; MLF1 adapter molecule; MUDPNP; multiple domain putative nuclear protein; myeloid leukemia factor 1 adaptor molecule; Nrbp; NRBP1; nuclear receptor binding protein 1; Nuclear receptor-binding protein; QtsA-11263; unnamed protein product | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29959 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UHY1 | |
| NRBP1 | |
| Recombinant Human Nuclear receptor-binding protein (406-535AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu