Learn More
NSE gamma Mouse, Clone: SPM347, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio 2026-MSM1X-P1
Code nomenclature Nacres: NA.46
Code produit. 15979947
  • ONLINE EXCLUSIVE PRICE
    554.40€
    478.00€ / 100µg
    Économisez 76.40€
    13% de réduction
Spécification
NSE gamma
Cytoplasmic
SPM347
Unconjugated
NP_001966
Mouse
50kDa
Protein A/G Purified
RUO
2026
IgG2b
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
ENO2
A synthetic peptide corresponding to aa416-433 of human NSE gamma
HepG2, SH-SY-5Y, HeLa or Y79 cells. Pancreas, Cerebellum or Pheochromocytoma.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.