missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NXN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554825
This item is not returnable.
View return policy
Description
Immunogen sequence: PIAEKIIAKY KAKEEEAPLL FFVAGEDDMT DSLRDYTNLP EAAPLLTILD MSARAKYVMD VEEITPAIVE AFVNDFLAEK LKPEPI Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
This gene encodes a member of the thioredoxin superfamily, a group of small, multifunctional redox-active proteins. Members of this family are characterized by a conserved active motif called the thioredoxin fold that catalyzes disulfide bond formation and isomerization. The encoded protein acts a redox-dependent regulator of the Wnt signaling pathway and is involved in cell growth and differentiation.
Specifications
| NXN | |
| Polyclonal | |
| Unconjugated | |
| NXN | |
| Gn25; l11Jus13; NRX; nucleoredoxin; nucleoredoxin 1; NXN; Protein Red-1; TRG-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18230, 360577, 64359 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P97346, Q6DKJ4 | |
| NXN | |
| Recombinant protein corresponding to Human NXN. Recombinant protein control fragment (Product #RP-93378). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction