Learn More
Invitrogen™ Onartuzumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA558284
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Onartuzumab has been used in trials studying the treatment of Neoplasms, Lung Cancer, Glioblastoma, Gastric Cancer, and Colorectal Cancer, among others.
Spécification
| Onartuzumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MetMAb; OA-5D5; PRO 143966 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |