missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR6A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5102216
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody detects endogenous levels of total OR6A2.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Spécification
| OR6A2 | |
| Polyclonal | |
| Unconjugated | |
| OR6A2 | |
| hI7 olfactory receptor; hP2 olfactory receptor; I7; Olfactory receptor 11-55; olfactory receptor 6A1; Olfactory receptor 6A2; olfactory receptor family 6 subfamily A member 2; olfactory receptor OR11-83; olfactory receptor, family 6, subfamily A, member 1; olfactory receptor, family 6, subfamily A, member 2 pseudogene; OR11-55; OR6A1; OR6A2; OR6A2P; seven transmembrane helix receptor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8590 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95222 | |
| OR6A2 | |
| A synthesized peptide derived from human OR6A2(Accession O95222), corresponding to amino acid residues N66-E116. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu