missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORP5 Polyclonal Antibody

Goat Polyclonal Antibody
Marque: Invitrogen™ PA518221
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors that play a key role in the maintenance of cholesterol balance in the body. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. This gene has been shown to be imprinted, with preferential expression from the maternal allele only in placenta. Transcript variants encoding different isoforms have been identified.
Spécification
| ORP5 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL5 | |
| DKFZp586H1719; FLJ31948; FLJ42929; KIAA1534; OBPH1; ORP5; ORP-5; OSBPL5; OSBP-related protein 5; oxysterol binding protein like 5; oxysterol binding protein-like 5; oxysterol-binding protein homolog 1; oxysterol-binding protein homologue 1; oxysterol-binding protein-related protein 5 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 114879, 361686 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9H0X9 | |
| OSBPL5 | |
| Synthetic peptide sequence (KEEAFLRRRFSLC) corresponding to the N-terminus amino acids of OSBPL5 (aa 2-15). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu