missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40 Ligand/TNFSF4 Antibody (R4930 (Oxelumab)), Allophycocyanin, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals NBP2-75913APC
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry.
Spécification
| OX40 Ligand/TNFSF4 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| TNFSF4 | |
| Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells. | |
| 0.1 mL | |
| Primary | |
| Binds to human OX40L. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| R4930 (Oxelumab) | |
| Flow Cytometry | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| Human | |
| Protein A purified | |
| Cancer, Cell Cycle and Replication, Immunology | |
| 7292.0 | |
| Human | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu