missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
P2Y11/P2RY11 Polyclonal antibody specifically detects P2Y11/P2RY11 in Human, Equine samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spécification
Spécification
| Antigène | P2Y11/P2RY11 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugué | Unconjugated |
| Dilution | ELISA 1:100 - 1:2000, Immunohistochemistry 8 μg/mL, Immunohistochemistry-Paraffin 8 μg/mL |
| Formule | PBS |
| Alias de gène | P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11 |
| Espèces hôtes | Rabbit |
| Immunogène | Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human P2Y11/P2RY11. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?