Learn More
p53 Antibody (PAb 240), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB200-103APC
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
p53 Monoclonal specifically detects p53 in Human, Mouse, Rat, Yeast, Xenopus (Negative) samples. It is validated for Flow Cytometry.
Spécification
| p53 | |
| Monoclonal | |
| APC | |
| Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This monoclonal recognizes both mutant forms and wild-type human p53 under denaturing conditions. It does not recognize nondenatured wild type p53. In nondenaturing conditions (IP) it only recognizes mutant p53. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| PAb 240 | |
| Flow Cytometry | |
| TP53 | |
| Gel-purified p53-beta-galactosidase fusion protein containing murine p53 from aa 14-389 (derived from pSV53C cDNA clone). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
| 7157 | |
| Human, Mouse, Rat, Yeast, Xenopus (Negative) | |
| Purified |
Correction du contenu d'un produit
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu