Learn More
pan Cytokeratin (pan CK) Mouse anti-Human, Clone: AE1+AE3, liquid, TrueMAB™
Mouse Monoclonal Antibody
Marque: Origene Technologies TA190321
Description
Pan cytokeratin protein include type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells. The product of this gene typically dimerizes with keratin 18 to form an intermediate filament in simple single-layered epithelial cells. This protein plays a role in maintaining cellular structural integrity and also functions in signal transduction and cellular differentiation. Mutations in this gene cause cryptogenic cirrhosis. Alternatively spliced transcript variants have been found for this gene.Spécification
| pan Cytokeratin (pan CK) | |
| Monoclonal | |
| Unconjugated | |
| KRT6A | |
| pan Cytokeratin | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3848, 3849, 3850, 3851, 3852, 3853, 3855, 3856, 3858, 3860, 3861, 3866, 3868, 3875, 3880 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| AE1+AE3 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P02533, P02538, P04264, P05783, P05787, P08727, P08729, P08779, P12035, P13645, P13646, P13647, P19012, P19013, P35908 | |
| KRT1, KRT10, KRT13, KRT14, KRT15, KRT16, KRT18, KRT19, KRT2, KRT3, KRT4, KRT5, KRT6A, KRT7, KRT8 | |
| Human epidermal cytokeratins | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.